4-(4-Fluorobenzyloxy)benzaldehyde 97%

Supplier: Apollo Scientific
Warning

Synonyms: 4-((4-Fluorobenzyl)oxy)benzaldehyde

PC9440-25G PC9440-5G
APOSPC9440-25GEA 382 EUR
APOSPC9440-25G APOSPC9440-5G
4-(4-Fluorobenzyloxy)benzaldehyde 97%
4-(4-Fluorobenzyloxy)benzaldehyde
Formula: C₁₄H₁₁FO₂
MW: 230,24 g/mol
Melting Pt: 96…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00173650
CAS Number: 56442-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR