7-Fluoroindole 98%
Supplier: Apollo Scientific
|
Warning
|
PC9454-1G
PC9454-5G
PC9454-25G
APOSPC9454-1GEA
23.2
EUR
APOSPC9454-1G
APOSPC9454-5G
APOSPC9454-25G
7-Fluoroindole 98%
7-Fluoroindole
Formula:
C₈H₆FN MW: 135,14 g/mol Melting Pt: 60…65 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01074502 CAS Number: 387-44-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...