2-(3-Chloro-4-fluorophenyl)ethanol 97%

Supplier: Apollo Scientific
Warning

Synonyms: 3-Chloro-4-fluorophenethyl alcohol

PC9510-1G PC9510-5G PC9510-25G
APOSPC9510-1GEA 55.1 EUR
APOSPC9510-1G APOSPC9510-5G APOSPC9510-25G
2-(3-Chloro-4-fluorophenyl)ethanol 97%
2-(3-Chloro-4-fluorophenyl)ethanol
Formula: C₈H₈ClFO
MW: 174,60 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06201121
CAS Number: 340825-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR