2-Iodo-4-(trifluoromethoxy)aniline

Supplier: Apollo Scientific
Danger

Synonyms: 4-(Trifluoromethoxy)-2-iodoaniline

PC9571-1G PC9571-250MG
APOSPC9571-1GEA 80 EUR
APOSPC9571-1G APOSPC9571-250MG
2-Iodo-4-(trifluoromethoxy)aniline
2-Iodo-4-(trifluoromethoxy)aniline
Formula: C₇H₅F₃INO
Storage Temperature: Ambient
MDL Number: MFCD04972994
CAS Number: 845866-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR