5-Fluoro-2-iodotoluene

Supplier: Apollo Scientific
Warning

PC9595-100G PC9595-500G
APOSPC9595-100GEA 80 EUR
APOSPC9595-100G APOSPC9595-500G
5-Fluoro-2-iodotoluene
5-Fluoro-2-iodotoluene
Formula: C₇H₆FI
MW: 236,03 g/mol
Boiling Pt: 138…139 °C
Storage Temperature: Ambient
MDL Number: MFCD00039397
CAS Number: 66256-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR