α-Chloro-2,3,4,5-tetrafluorotoulene
Supplier: Apollo Scientific
|
Danger
|
PC9598-1G
PC9598-5G
APOSPC9598-1GEA
40.6
EUR
APOSPC9598-1G
APOSPC9598-5G
α-Chloro-2,3,4,5-tetrafluorotoulene
α-Chloro-2,3,4,5-tetrafluorotoulene
Formula:
C₇H₃ClF₄ Boiling Pt: 125 °C/7mmHg Storage Temperature: Refrigerator |
CAS Number:
21622-18-4 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...