Methyl-2,5-bis(trifluoroethoxy)benzoate 95+%
Supplier: Apollo Scientific
|
Warning
|
PC9659-10G
PC9659-1G
PC9659-5G
APOSPC9659-10GEA
87.3
EUR
APOSPC9659-10G
APOSPC9659-1G
APOSPC9659-5G
Methyl-2,5-bis(trifluoroethoxy)benzoate 95+%
Methyl-2,5-bis(trifluoroethoxy)benzoate
Formula:
C₁₂H₁₀F₆O₄ MW: 332,2 g/mol Melting Pt: 40…41 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236693 CAS Number: 35480-31-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...