2-Nitro-α,α,α-trifluoro-p-toluic acid 97%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Nitro-4-(trifluoromethyl)benzoic acid, 2-Nitro-4-trifluoromethylbenzoic acid

PC9666-25G PC9666-5G PC9666-100G
APOSPC9666-25GEA 27.7 EUR
APOSPC9666-25G APOSPC9666-5G APOSPC9666-100G
2-Nitro-α,α,α-trifluoro-p-toluic acid 97%
2-Nitro-α,α,α-trifluoro-p-toluic acid
Formula: C₈H₄F₃NO₄
MW: 235,12 g/mol
Melting Pt: 134…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00007142
CAS Number: 320-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR