2-(3-Bromo-4-fluorophenyl)acetonitrile 95%

Supplier: Apollo Scientific
Warning

Synonyms: 3-Bromo-4-fluorophenylacetonitrile

PC9669-10G PC9669-25G PC9669-5G
APOSPC9669-10GEA 133 EUR
APOSPC9669-10G APOSPC9669-25G APOSPC9669-5G
2-(3-Bromo-4-fluorophenyl)acetonitrile 95%
2-(3-Bromo-4-fluorophenyl)acetonitrile
Formula: C₈H₅BrFN
MW: 214,037 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08458154
CAS Number: 501420-63-9
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR