6-chloro-5-fluoroindole
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
6-Chloro-5-fluoroindole
PC9724-1G
PC9724-250MG
APOSPC9724-1GEA
39.3
EUR
APOSPC9724-1G
APOSPC9724-250MG
6-chloro-5-fluoroindole
6-chloro-5-fluoroindole
Formula:
C₈H₅ClFN Melting Pt: 104…107 °C Storage Temperature: Ambient |
MDL Number:
MFCD01074496 CAS Number: 122509-72-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...