4,4,4-Trifluorobutylamine 97%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
1-Butanamine,4,4,4-trifluoro-, 4,4,4-trifluorobutan-1-amine, 1-Amino-4,4,4-trifluorobutane
PC9728-1G
PC9728-5G
APOSPC9728-1GEA
52.2
EUR
APOSPC9728-1G
APOSPC9728-5G
4,4,4-Trifluorobutylamine 97%
4,4,4-Trifluorobutylamine
Formula:
C₄H₈F₃N MW: 127,11 g/mol Boiling Pt: 90…94 °C Density: 1,35 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD06213030 CAS Number: 819-46-5 UN: 2734 ADR: 8+3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...