6-Fluoroindol-3-ylacetic acid

Supplier: Apollo Scientific
Warning

PC9731-1G PC9731-500MG PC9731-5G
APOSPC9731-1GEA 176 EUR
APOSPC9731-1G APOSPC9731-500MG APOSPC9731-5G
6-Fluoroindol-3-ylacetic acid
6-Fluoroindol-3-ylacetic acid
Formula: C₁₀H₈FNO₂
MW: 193,18 g/mol
Melting Pt: 159…163 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01074503
CAS Number: 443-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR