5-Ethynyl-1,2,3-trifluorobenzene

Supplier: Apollo Scientific
Warning

PC9819-1G PC9819-5G PC9819-25G
APOSPC9819-1GEA 66.7 EUR
APOSPC9819-1G APOSPC9819-5G APOSPC9819-25G
5-Ethynyl-1,2,3-trifluorobenzene
5-Ethynyl-1,2,3-trifluorobenzene
Formula: C₈H₃F₃
Boiling Pt: 53…54 °C/1mmHg
Storage Temperature: Ambient
MDL Number: MFCD08458177
CAS Number: 158816-55-8
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR