1-Fluorobutane
Supplier: Apollo Scientific
|
Danger
|
PC9870-25G
PC9870-5G
APOSPC9870-25GEA
510
EUR
APOSPC9870-25G
APOSPC9870-5G
1-Fluorobutane
1-Fluorobutane
Formula:
C₄H₉F MW: 76.11 g/mol Boiling Pt: 32 °C/745mmHg Density: 0.776 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00039288 CAS Number: 2366-52-1 UN: 1993 ADR: 3 I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...