2-Chloro-5-(trifluoromethyl)-4-iodopyridine

Supplier: Apollo Scientific
Warning

Synonyms: 2-Chloro-4-iodo-5-(trifluoromethyl)pyridine

PC9878-1G PC9878-25G PC9878-5G
APOSPC9878-1GEA 19 EUR
APOSPC9878-1G APOSPC9878-25G APOSPC9878-5G
2-Chloro-5-(trifluoromethyl)-4-iodopyridine
2-Chloro-5-(trifluoromethyl)-4-iodopyridine
Formula: C₆H₂ClF₃IN
MW: 307,44 g/mol
Melting Pt: 126…127 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01862099
CAS Number: 505084-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR