Bis(2,2,3,3,3-pentafluoropropyl)amine 95%

Supplier: Apollo Scientific
Warning

PC9891-1G PC9891-25G PC9891-5G
APOSPC9891-1GEA 109 EUR
APOSPC9891-1G APOSPC9891-25G APOSPC9891-5G
Bis(2,2,3,3,3-pentafluoropropyl)amine 95%
Bis(2,2,3,3,3-pentafluoropropyl)amine
Formula: C₆H₅F₁₀N
MW: 281,1 g/mol
Boiling Pt: 101…103 °C
Storage Temperature: Ambient
MDL Number: MFCD04038263
CAS Number: 883498-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR