3-Fluoro-2-hydroxypyridine
Supplier: Apollo Scientific
|
Warning
|
PC9928-1G
PC9928-5G
PC9928-25G
APOSPC9928-1GEA
21.9
EUR
APOSPC9928-1G
APOSPC9928-5G
APOSPC9928-25G
3-Fluoro-2-hydroxypyridine
3-Fluoro-2-hydroxypyridine
Formula:
C₅H₄FNO MW: 113,09 g/mol Melting Pt: 157…161 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095042 CAS Number: 1547-29-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...