1-Bromo-2-fluoro-5-nitrobenzene 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Bromo-4-fluoronitrobenzene, 2-Bromo-1-fluoro-4-nitrobenzene
PC9956-100G
PC9956-25G
PC9956-5G
APOSPC9956-100GEA
48
EUR
APOSPC9956-100G
APOSPC9956-25G
APOSPC9956-5G
1-Bromo-2-fluoro-5-nitrobenzene 98%
1-Bromo-2-fluoro-5-nitrobenzene
Formula:
C₆H₃BrFNO₂ MW: 220 g/mol Melting Pt: 55…58 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00055433 CAS Number: 701-45-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...