2-Methylsulphonyl-1-[6-(trifluoromethyl)pyridin-3-yl]ethanone 95+%

Supplier: Apollo Scientific
Warning

Synonyms: 5-[(Methylsulphonyl)acetyl]-2-(trifluoromethyl)pyridine

PC9974-1G PC9974-250MG
APOSPC9974-1GEA 256 EUR
APOSPC9974-1G APOSPC9974-250MG
2-Methylsulphonyl-1-[6-(trifluoromethyl)pyridin-3-yl]ethanone 95+%
2-Methylsulphonyl-1-[6-(trifluoromethyl)pyridin-3-yl]ethanone
Formula: C₉H₈F₃NO₃S
MW: 267,23 g/mol
Melting Pt: 129…131 °C
Storage Temperature: Ambient
MDL Number: MFCD01862655
CAS Number: 386704-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR