2',6'-Difluoroacetanilide
Supplier: Apollo Scientific
Synonyms:
N-(2,6-Difluorophenyl)acetamide
PC9976-1G
PC9976-5G
APOSPC9976-1GEA
109
EUR
APOSPC9976-1G
APOSPC9976-5G
2',6'-Difluoroacetanilide
2',6'-Difluoroacetanilide
Formula:
C₈H₇F₂NO MW: 171,144 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00119485 CAS Number: 3896-29-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...