3-Chloroheptafluorobut-2-ene 98%

Supplier: Apollo Scientific
Warning

PC9981-100G PC9981-25G
APOSPC9981-100GEA 430 EUR
APOSPC9981-100G APOSPC9981-25G
3-Chloroheptafluorobut-2-ene 98%
3-Chloroheptafluorobut-2-ene
Formula: C₄ClF₇
MW: 216,49 g/mol
Boiling Pt: 32 °C
Density: 1,5482 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00039261
CAS Number: 434-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR