Phenol : chloroform : iso-amyl alcohol 25:24:1 stabilised

Supplier: Apollo Scientific

Synonyms: Carbolic acid, Hydroxybenzene, Phenic acid

Stabilisation: Stabilised with 8-hydroxyquinoline 0,1 %

BIA0889-100ML BIA0889-500ML
APOSBIA0889-100MLEA 94.5 EUR
APOSBIA0889-100ML APOSBIA0889-500ML
Phenol : chloroform : iso-amyl alcohol 25:24:1 stabilised
Phenol : Chloroform : iso-Amyl alcohol

Phenol equilibrated, stabilised:chloroform :isoamyl alcohol 25:24:1


Caution: Shelf life approx. 9 months

Density: 1,280 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00133763
CAS Number: 136112-00-0
UN: 2922
ADR: 8+6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR