CryoBoxes, Cardboard, Origami
Supplier: TENAK
TE22825
479-1175EA 270 EUR
479-1175
CryoBoxes, Cardboard, Origami
Boxes Cryoboxes
Cryoboxes, white cardboard, PP coated.
  • PP coated for longer durability
  • Easy to assemble
  • Water-repellant
  • Delivered 'flat-packed' and unassembled: Saves shipping cost and storage space in the laboratory