Valsartan
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(S)-2-(N-((2'-(1H-Tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)pentanamido)-3-methylbutanoic acid, (S)-2-(N-((2'-(1H-Tetrazol-5-yl)[1,1'-biphenyl]-4-yl)methyl)pentanamido)-3-methylbutanoic acid
465560010
465560050
ACRO465560010EA
44
EUR
ACRO465560010
ACRO465560050
Valsartan
Valsartan
Formula:
C₂₄H₂₉N₅O₃ MW: 435,52 g/mol Melting Pt: 117°C Storage Temperature: Refrigerator |
MDL Number:
MFCD00865840 CAS Number: 137862-53-4 Merck Index: 13.9982 |
Specification Test Results
Appearance | White to almost white to light beige powder |
Infrared spectrum | Conforms |
HPLC | ≥ 97.5% |
Specific optical rotation | -55° to -70° (20°C, 589nm) (1% in CH3OH) |
Water | ≤ 2% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...