Latanoprost
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Latanoprost, (Z)-Isopropyl 7-((1R,2R,3R,5S)-3,5-dihydroxy-2-((R)-3-hydroxy-5-phenylpentyl)cyclopentyl)hept-5-enoate
466660250
466661000
ACRO466660250EA
158
EUR
ACRO466660250
ACRO466661000
Latanoprost
Latanoprost
Formula:
C₂₆H₄₀O₅ MW: 432,60 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00216074 CAS Number: 130209-82-4 UN: 2810 ADR: 6.1,III Merck Index: 13.5391 |
Specification Test Results
Appearance | Clear colorless to light yellow liquid |
Infrared spectrum | Conforms |
HPLC | ≥ 98.5% |
Water | ≤ 0.5% |
Residue after ignition | ≤ 0.5% |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...