Premium syringe barrels and end caps, 700 system
Supplier: TECHCON SYSTEMS INC
7030LL1NPK 7050LL1NPK 7100LL1BPK
TESI7030LL1NPKEA 0 EUR
TESI7030LL1NPK TESI7050LL1NPK TESI7100LL1BPK
Premium syringe barrels and end caps, 700 system
Syringe Barrels, Plungers and Pistons
Premium syringe barrels are available in four colours.
Natural colour is the industry standard. Dark amber colour ensures UV and visible light blocking plus the ability to see inside the barrel.
Carbon-free black colour prevents carbon contamination while providing maximum protection from UV and visible light. Red end caps also block UV light.