4,5-Dimethoxy-2-nitrobenzamide ≥96%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Carbamoyl-4,5-dimethoxynitrobenzene

OR8586-1G
APOSOR8586-1GEA 0 EUR
APOSOR8586-1G
4,5-Dimethoxy-2-nitrobenzamide ≥96%
4,5-Dimethoxy-2-nitrobenzamide
Formula: C₉H₁₀N₂O₅
MW: 226,19 g/mol
MDL Number: MFCD00171550
CAS Number: 4959-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR