Antimicrobial Trays and Inserts
Supplier: GRATNELL'S
FA0120P12 FA0129P12 FA2529P6
GRATFA0120P12EA 299 EUR
GRATFA0120P12 GRATFA0129P12 GRATFA2529P6
Antimicrobial Trays and Inserts
Trays/Dishes/Pots Laboratory Trays/Dishes
The Gratnells Antimicrobial trays are treated with BioCote® antibacterial additive. Seamlessly integrates with standard school furniture. Available in 4 heights.
  • BSI tested and passed for heavy educational use
  • Fully recyclable
  • Lids and inserts available
  • Scratch resistant
  • Anti-static additive-protection from dust contamination
  • Anti-fade colour protector
When conducting a mini-experiment or demonstration, the tray safely keeps all chemicals contained and prevents contamination of the work surface. Useful for storing chemicals on shelving ensuring any leakages are contained. Students can select pre-prepared resources from trays, taking responsibility for their learning. Equipment can be sorted and transported around the learning space with ease, preventing lost learning time.
Chemically unreactive polypropylene & Semi fire-retardant at 0,22 mm per min.