Gibberellic acid

Supplier: Roth Carl

Synonyms: GA3, Gibberellin A3, (3S,3aS,4S,4aS,6S,8aR,8bR,11S)-6,11-Dihydroxy-3-methyl-12-methylene-2-oxo-4a,6-ethano-3,8b-prop-1-enoperhydroindeno[1,2-b]furan-4-carboxylic acid

7464.1 7464.2
ROTH7464.1EA 0 EUR
ROTH7464.1 ROTH7464.2
Gibberellic acid
Gibberellic acid
Formula: C₁₉H₂₂O₆
MW: 346.38 g/mol
Melting Pt: 234
Density: 1.386 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00079329
CAS Number: 77-06-5
EINECS: 201-001-0
Merck Index: 13,04430

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR