(±)-4-Hydroxy-3-methoxymandelic acid ≥98%

Supplier: Thermo Fisher Scientific
Warning

A13349.14 A13349.06 A13349.03
A13349.14EA 881 EUR
A13349.14 A13349.06 A13349.03
(±)-4-Hydroxy-3-methoxymandelic acid ≥98%
(±)-4-Hydroxy-3-methoxymandelic acid

Purity: 98% CAS No.: 2394-20-9 Molecular Formula: C9H10O5 Formula Weight: 198.17 Melting Point: 133-134° dec. MDL No.: MFCD00004235

Formula: C₉H₁₀O₅
Boiling Pt: 421 °C (1013 hPa)
Melting Pt: 133…134 °C (dec.)
Density: 1,44 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004235
CAS Number: 55-10-7
EINECS: 200-224-0
Merck Index: 12,10070

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR