Dibutyl sulphate 95%

Supplier: Apollo Scientific
Danger

OR924973-5G OR924973-25G OR924973-100G
APOSOR924973-5GEA 72.5 EUR
APOSOR924973-5G APOSOR924973-25G APOSOR924973-100G
Dibutyl sulphate 95%
Dibutyl sulphate
Formula: C₈H₁₈O₄S
Boiling Pt: 115…116 °C
Density: 1,06
MDL Number: MFCD00059420
CAS Number: 625-22-9
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR