Dimethylmalononitrile

Supplier: Apollo Scientific
Danger

OR71123-5G OR71123-25G OR71123-100G
APOSOR71123-5GEA 97.3 EUR
APOSOR71123-5G APOSOR71123-25G APOSOR71123-100G
Dimethylmalononitrile
Dimethylmalononitrile
Formula: C₅H₆N₂
Melting Pt: 31 - 33 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01321253
CAS Number: 7321-55-3
UN: 2926
ADR: 4.1+6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR