Butyl oleate 95%
Supplier: Apollo Scientific
|
Warning
|
OR918733-25G
OR918733-5G
APOSOR918733-25GEA
43.5
EUR
APOSOR918733-25G
APOSOR918733-5G
Butyl oleate 95%
Butyl oleate
Formula:
C₂₂H₄₂O₂ |
MDL Number:
MFCD00053819 CAS Number: 142-77-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...