Haematoxylin

Supplier: Roth Carl

Synonyms: DcH-48, Hydroxybrazilin, (6aS,11bR)-6,6a,7,11b-Tetrahydroindeno[2,1-c]chromene-3,4,6a,9,10-pentaol, NSC 2785, Natural Black I, Hydroxybrasilin, (+)-Hematoxylin

3816.1 3816.2
ROTH3816.1EA 80.4 EUR
ROTH3816.1 ROTH3816.2
Haematoxylin
Haematoxylin
Formula: C₁₆H₁₄O₆
MW: 302,28 g/mol
Melting Pt: 291…293 °C
Storage Temperature: Ambient
MDL Number: MFCD00078111
CAS Number: 517-28-2
EINECS: 208-237-3
Merck Index: 13,04654

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR