2-Amino-4-methylpyrimidine 97%

Supplier: Thermo Scientific
Warning

Synonyms: 4-Methylpyrimidin-2-ylamine, 4-Methylpyrimidin-2-amine

104090100 104091000
ACRO104090100EA 30 EUR
ACRO104090100 ACRO104091000
2-Amino-4-methylpyrimidine 97%
2-Amino-4-methylpyrimidine
Formula: C₅H₇N₃
MW: 109,13 g/mol
Boiling Pt: 283 °C (1013 hPa)
Melting Pt: 155…160 °C
Density: 1 g/cm³ (20 °C)
MDL Number: MFCD00006101
CAS Number: 108-52-1
EINECS: 203-591-5

Order Now

Specification Test Results

Appearance Off-white to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 155 to 160 °C
Titration with HClO4 ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR