(±)-γ-Valerolactone 98%
Supplier: Thermo Fisher Scientific
Synonyms:
(±)-4-Hydroxypentanoic acid lactone, (±)-Pentan-4-olide, γ-Pentanolactone, 5-Methyldihydrofuran-2(3H)-one, gamma-Valerolactone
140791000
140795000
ACRO140791000EA
64.7
EUR
ACRO140791000
ACRO140795000
(±)-γ-Valerolactone 98%
(±)-γ-Valerolactone
Formula:
C₅H₈O₂ Boiling Pt: 207…208 °C (760 mmHg) Melting Pt: –31 °C Density: 1,05 g/cm³ Flash Pt: 96 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005400 CAS Number: 108-29-2 EINECS: 203-569-5 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4310 to 1.4340 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...