Methyl cyanoacetate 97%
Supplier: Thermo Scientific
Synonyms:
Methyl 2-cyanoacetate
148640010
148642500
ACRO148640010EA
120
EUR
ACRO148640010
ACRO148642500
Methyl cyanoacetate 97%
Methyl cyanoacetate
Formula:
C₄H₅NO₂ MW: 99,09 g/mol Boiling Pt: 207 °C (1013 hPa) Melting Pt: –13 °C Density: 1,12 g/cm³ Flash Pt: >110 °C |
MDL Number:
MFCD00001939 CAS Number: 105-34-0 EINECS: 203-288-8 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4170 to 1.4190 (20°C, 589 nm) |
GC | ≥96.0 % |
Water | ≤5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...