Crotonic acid 98%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
trans-Crotonic acid, 2-Butenoic acid, trans-2-Butenoic acid
150875000
150870025
ACRO150875000EA
56.8
EUR
ACRO150875000
ACRO150870025
Crotonic acid 98%
Crotonic acid
Formula:
C₄H₆O₂ Boiling Pt: 185 °C (760 mmHg) Melting Pt: 72 °C Density: 1,02 g/cm³ (20 °C) Flash Pt: 88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002701 CAS Number: 107-93-7 EINECS: 203-533-9 UN: 2823 ADR: 8,III Merck Index: 13,02625 |
Specification Test Results
Appearance | White to light yellow Crystalline flakes |
Infrared spectrum | Conforms |
Water | ≤0.7 % |
GC | ≥97.5 % |
Titration with NaOH | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...