3-Nitrophthalic acid 95%
Supplier: Thermo Fisher Scientific
|
Warning
|
160161000
160165000
160160025
ACRO160161000EA
61.8
EUR
ACRO160161000
ACRO160165000
ACRO160160025
3-Nitrophthalic acid 95%
3-Nitrophthalic acid
Formula:
C₈H₅NO₆ MW: 211.13 g/mol Boiling Pt: 441 Melting Pt: 213…216 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00007138 CAS Number: 603-11-2 EINECS: 210-030-8 |
Specification Test Results
Appearance | Off-white to light yellow Powder |
Infrared spectrum | Conforms |
HPLC | ≥94 % |
Titration with NaOH | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...