α-Bromo-o-toluonitrile 98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2-Cyanobenzyl bromide, 2-(Bromomethyl)benzonitrile, o-(Bromomethyl)benzonitrile
163130250
163131000
163130050
ACRO163130250EA
133
EUR
ACRO163130250
ACRO163131000
ACRO163130050
α-Bromo-o-toluonitrile 98%
α-Bromo-o-toluonitrile
Formula:
C₈H₆BrN MW: 196,05 g/mol Boiling Pt: 152…155 °C (15 mmHg) Melting Pt: 70…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001794 CAS Number: 22115-41-9 UN: 1759 ADR: 8,II |
Specification Test Results
Appearance | White to grey-brown Crystalline powder and/or chunks |
Infrared spectrum | Conforms |
Melting point | 70 to 74 °C |
GC | ≥97.5 % |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...