Methyl-3,5-dihydroxybenzoate 97%

Supplier: Thermo Scientific
Warning

Synonyms: Methyl 3,5-dihydroxybenzoate, 3,5-Dihydroxybenzoic acid methyl ester

170440250 170441000
ACRO170440250EA 96.7 EUR
ACRO170440250 ACRO170441000
Methyl-3,5-dihydroxybenzoate 97%
Methyl 3,5-dihydroxybenzoate
Formula: C₈H₈O₄
MW: 168,15 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 164…169 °C
MDL Number: MFCD00002289
CAS Number: 2150-44-9
EINECS: 218-426-2

Order Now

Specification Test Results

Appearance Light yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 164 to 169 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR