5-Amino-4,6-dichloropyrimidine 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
4,6-Dichloropyrimidin-5-ylamine
203250050
203250250
ACRO203250050EA
174
EUR
ACRO203250050
ACRO203250250
5-Amino-4,6-dichloropyrimidine 97%
5-Amino-4,6-dichloropyrimidine
Formula:
C₄H₃Cl₂N₃ Boiling Pt: 262 °C (1013 hPa) Melting Pt: 143…148 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006108 CAS Number: 5413-85-4 EINECS: 226-503-7 |
Specification Test Results
Appearance | Yellow-brown Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 143 to 148 °C |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...