3-Thiopheneethanol 99%

Supplier: Thermo Scientific

Synonyms: Thiophen-3-ethanol, 2-(Thiophen-3-yl)ethanol, 2-(3-Thienyl)ethanol

211710050 211710250
ACRO211710050EA 95.4 EUR
ACRO211710050 ACRO211710250
3-Thiopheneethanol 99%
3-Thiopheneethanol
Formula: C₆H₈OS
MW: 128,19 g/mol
Boiling Pt: 110…111 °C (14 mmHg)
Density: 1,14 g/cm³
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00009766
CAS Number: 13781-67-4
EINECS: 237-434-7

Order Now

Specification Test Results

Appearance Clear colorless to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.5510 to 1.5550 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR