3-Formylbenzoic acid 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Isophthalaldehydic acid, 3-Carboxybenzaldehyde
296560010
296560050
ACRO296560010EA
59
EUR
ACRO296560010
ACRO296560050
3-Formylbenzoic acid 98%
3-Formylbenzoic acid
Formula:
C₈H₆O₃ MW: 150,13 g/mol Boiling Pt: 306 °C (1013 hPa) Melting Pt: 172…176 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039575 CAS Number: 619-21-6 EINECS: 210-585-6 |
Specification Test Results
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
Melting point | 172 to 176 °C |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...