N-Hydroxyphthalimide 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Hydroxyisoindoline-1,3-dione, 2-Hydroxy-1H-isoindole-1,3(2H)-dione, N-Hydroxypthalimide
329871000
329875000
329870025
ACRO329871000EA
42.4
EUR
ACRO329871000
ACRO329875000
ACRO329870025
N-Hydroxyphthalimide 98%
N-Hydroxyphthalimide
Formula:
C₈H₅NO₃ MW: 163,13 g/mol Boiling Pt: 412 °C (1013 hPa) Melting Pt: 233…235 °C Density: 1,58 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00005891 CAS Number: 524-38-9 EINECS: 208-358-1 Merck Index: 13,04864 |
Specification Test Results
Appearance | Yellow Powder |
Infrared spectrum | Conforms |
Water | ≤2 % |
Titration with NaOH | 97.5 to 102.5 % (On anhydrous substance) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...