Benzyltrimethylammonium bromide 98+%

Supplier: Thermo Scientific
Warning

Synonyms: N,N,N-Trimethyl-1-phenylmethanaminium bromide

364860250 364861000
ACRO364860250EA 26.8 EUR
ACRO364860250 ACRO364861000
Benzyltrimethylammonium bromide 98+%
Benzyltrimethylammonium bromide
Formula: C₁₀H₁₆BrN
MW: 230,15 g/mol
Melting Pt: 228…234 °C
MDL Number: MFCD00011780
CAS Number: 5350-41-4
EINECS: 226-320-2
UN: 2811

Order Now

Specification Test Results

Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Melting point 224 to 234 °C
Titration Argentometric ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR