Formylhydrazine 90%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Formic acid hydrazide, Formohydrazide, Formic hydrazide

376060250 376061000
ACRO376060250EA 93.8 EUR
ACRO376060250 ACRO376061000
Formylhydrazine 90%
Formylhydrazine
Formula: CH₄N₂O
MW: 60.06 g/mol
Boiling Pt: 90 °C (0.7 mmHg)
Melting Pt: 51…58 °C
Flash Pt: 112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007589
CAS Number: 624-84-0
EINECS: 210-867-9

Order Now

Specification Test Results

Appearance White to pink Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 51 to 59 °C
Titration with KIO3 88 to 112 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR