Benzoylacetonitrile 98+%

Supplier: Thermo Scientific
Warning

Synonyms: 3-Phenyl-3-oxopropanenitrile, 3-Oxo-3-phenylpropanenitrile

402000050 402000250
ACRO402000050EA 70.8 EUR
ACRO402000050 ACRO402000250
Benzoylacetonitrile 98+%
Benzoylacetonitrile
Formula: (C₆H₅)C(O)CH₂CN
MW: 145,16 g/mol
Boiling Pt: 160 °C (10 mmHg)
Melting Pt: 78…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00001942
CAS Number: 614-16-4
EINECS: 210-369-1
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Light yellow to yellow-brown Crystalline powder
Infrared spectrum Conforms
Melting point 78 to 82 °C
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR