Sodium chloroacetate 98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Chloroacetic acid sodium salt

404415000 404410050 404410025
ACRO404415000EA 54.2 EUR
ACRO404415000 ACRO404410050 ACRO404410025
Sodium chloroacetate 98%
Sodium chloroacetate
Formula: C₂H₂ClNaO₂
Melting Pt: 199 °C
Storage Temperature: Ambient
MDL Number: MFCD00002684
CAS Number: 3926-62-3
EINECS: 223-498-3
UN: 2659
ADR: 6.1,III
Merck Index: 13,02129

Order Now

Specification Test Results

Appearance White Crystalline powder or granules or flakes
Infrared spectrum Conforms
Titration with HClO4 97.5 to 102.5 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR