Isoxazole 99%

Supplier: Thermo Scientific
Danger

412930100 412930500
ACRO412930100EA 80 EUR
ACRO412930100 ACRO412930500
Isoxazole 99%
Isoxazole
Formula: C₃H₃NO
Boiling Pt: 95 °C
Density: 1,07 g/cm³
Flash Pt: 8 °C
MDL Number: MFCD00003149
CAS Number: 288-14-2
EINECS: 206-018-7
UN: 1993

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4260 to 1.4280 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR